Burkert 3233 2-Way Diaphragm Valve, 316L SS and PTFE; 1/2" Tri-Clamp
Supplier: BURKERT FLUID CONTROL SYSTEMS
VWR Item#MFLX98516-60
Your Organization's Price:
Retrieving
Fluids are hermetically sealed from operating system.
  • Zero dead volume
  • Autoclavable
Choose from EPDM or PTFE wetted materials and sanitary or weld end connections. These compact, manually-operated diaphragm valves consist of a flow-optimized stainless steel body, diaphragm, and a manual actuator. The diaphragm is both a switch element and a sealing element and can be easily replaced. The valves have no dead volume and can be mounted to be self-draining — this enables high flow capacities and a variety of applications to be realized. The flow can be continually adjusted with the handwheel, making these valves extremely useful in applications in the food industry, biotechnology, and pharmaceutical markets.