Masterflex® Transfer Tubing, Cleargreen® PVC, 3/8" ID x 5/8" OD; 50 Ft
Supplier: VWR International
VWR Item#MFLX96605-33
Your Organization's Price:
Retrieving
Eliminate odor transferring to sample.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: ³/₈" int.Ø × ⁵/₈" out.Ø
<B>Characteristics:</B> Translucent, odorless, low water absorption, low extractables <br /><B>Certified compliant</B> FDA compliant, USP Class VI, 3A compliant, Reach compliant and RoHS compliant.<br /><B>Temperature range:</B> -58 to 165 °F (-50 to 74 °C) <br /><B>Sterilize:</B> Autoclave or ethylene oxide.