Masterflex® Transfer Tubing, PVC, 3/8" ID x 5/8" OD; 50 Ft
Supplier: SANISURE, INC. - SPARTA
VWR Item#MFLX96605-10
Your Organization's Price:
Retrieving
Stop oxidation and bacteria problems.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 3/8" ID x 5/8" OD
This tubing is extremely flexible and transparent and is resistant to oxidation, bacteria and a broad range of chemicals. PVC tubing meets FDA CFR Title 21 177.101.