ChemFluor PFA Tubing, 7/16" ID×1/2" OD; 50 ft/Pk
Supplier: VWR International
Meets the demands of various industries.
- Ideal for general-purpose, single-use, and extended fluid path applications
<B>Applications:</B>Pharmaceutical processes, cable covering, electrical applications.<br /><B>Characteristics:</B>Excellent temperature range, chemically inert, tough, flexible, low friction coefficient, Ozone-, UV-, and stress and crack resistant. Translucent.<br /><B>Certification:</B>FDA compliant. Meets USP Class VI requirements.<br /><B>Temperature range:</B>–320 to 500 °F (–195 to 260 °C).<br /><B>Sterilize:</B>With autoclave, ethylene oxide, or use dry heat.