Masterflex® Clamp Ties with Curved Head, White Plastic; 100/PK
Supplier: VWR International
Special curved clamp tie head ensures tight seal.
- Ideal for general-purpose, single-use applications
- Clamp size: 0.228" x 0.256"
Perfect for flexible fuel line, water line, and vacuum line applications. Ties measure 8" L x 0.18" W; maximum diameter of 1-3/4".