High-Density Polyethylene (HDPE) Tubing, 13/16" ID×1" OD; 100 Ft
Supplier: VWR International
VWR Item#MFLX95501-22
Your Organization's Price:
Retrieving
Designed for general purpose lab applications.
<B>Applications:</B> General-purpose lab and industrial use.<br /><B>Characteristics:</B> Low moisture absorption, good chemical and corrosion-resistance, high tensile strength.<br /><B>Certifications:</B> None.<br /><B>Temperature range:</B> –103 to 151 °F (–75 to 66 °C)<br /><B>Sterilize:</B> Autoclave.