High-pressure, brass-bodied rotary vane pump, 2.3 GPM, 115/230 VAC, 50/60 Hz
Supplier: BERNS CORP
VWR Item#MFLX79410-10
Your Organization's Price:
Retrieving
Ideal for high pressure applications—up to 240 psi.
  • Pulseless flow at pressures up to 240 psi
  • Built-in bypass relief valve protects your pump from damage
  • Select from economical brass or corrosion-resistant 303 stainless steel bodied pumps
Power cord not included.
115 VAC version only. Call our Application Specialists for 230 V version.