Masterflex® L/S® High-Performance Pump Head Replacement Mounting Kit
Supplier: Avantor Fluid Handling
Secure your Masterflex® High-Performance pump head!
- Constructed of durable stainless steel
Keep your pump head operating at its best with these replacement parts.
Delivery information: Includes four 10-24 stainless steel mounting screws.