Masterflex® Slide Forceps, plastic, wide tip; 50/pk
Supplier: VWR International
No more cross-contamination.
- Protect yourself and your samples with disposable forceps
- Autoclavable plastic or stainless steel construction
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.