High-Flow Flexible Impeller Pump, 8 GPM or 28 PSI, Neoprene Impeller, 3/4 HP
Supplier: BPH PUMP & EQUIPMENT INC
VWR Item#MFLX75501-12
Your Organization's Price:
Retrieving
Pumps fluids in applications where durability is needed—the 12 VDC models are ideal for remote/portable use.
  • Pharmaceutical—processing lab solutions, lotions, preparations, and filtering solutions
  • Industrial—circulating and transferring corrosive liquids, transferring solutions with entrained air (such as soaps and detergents), sampling, and pilot plant installations
  • Agricultural—transferring liquid fertilizers, herbicides, and pesticides
  • Metal plating—filtration of plating solutions, filling, and emptying vats
These flexible impeller self-priming non-metallic pumps are available in low-flow and high-flow options. <br /><br />Low-flow pumps are ideal for applications where gas-laden fluids need to be transferred. Built-in thermal overload protection prevents motor damage. Model 75421-00 is constructed with a flass-filled polyester body, Buna-N impeller, and stainless steel (SS) shaft. <br /><br />High-flow pumps are ideal for higher flow applications that require continuous duty. The epoxy body offers great resistance to a wide range of fluids; choose an impeller material compatible with your pumped fluid. Motors have a conduit box for easy wiring.
<B>High-Flow Pumps</B> are ideal for mid to higher flow applications. The epoxy body offers great resistance to a wide range of fluids; choose an impeller material compatible with your pumped fluid. Motors have a conduit box for easy wiring.