DC-Powered Diaphragm Pump; 12 VDC, Pulse
Supplier: BPH PUMP & EQUIPMENT INC
VWR Item#MFLX74201-01
Your Organization's Price:
Retrieving
DC-powered pumps with PVDF pump head.
  • High resolution with a flow range of 0.055 to 38 ml/min
  • Choose between 12 VDC or 24 VDC power—pulse, start/stop, or proportional control
  • Connect 1/8" x 1/4" tubing directly to pump ports
Choose a pump with one of the following control methods: <br />Pulse: 1 stroke per pulse—0 to 720 strokes per minute control capability<br />4 to 20 mA: proportional control based on mA signal<br />1 to 5 VDC: proportional control based upon a 1 to 5 VDC input signal<br />Start/stop: pump stops during contact input