Masterflex® Power Cord, 230 VAC, Indian Plug; 6-ft Long
Supplier: VWR International
VWR Item#MFLX50001-64
Your Organization's Price:
Retrieving
Optimize performance of your Masterflex® pump.
  • Choose from a wide range of power cords
Choose an interchangeable cord/plug assembly for 115 or 230 VAC operation. Each 6-ft (1.8 m) cord and moded plug assembly features an IEC 320 appliance connector and rated for 10 A at 250 VAC.