Masterflex® Fitting, Polypropylene, Straight, Female Luer to Hosebarb Adapters, 1/16"; 25/PK
Supplier: VWR International
VWR Item#MFLX45508-00
Your Organization's Price:
Retrieving
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female Luer to Hose Barb, 1/16"