Masterflex® Fitting, Nylon, Bulkhead, Female Luer to Hose Barb Adapter, 1/8" ID; 25/PK
Supplier: VWR International
VWR Item#MFLX45502-34
Your Organization's Price:
Retrieving
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female Luer x 1/4-28 UNF to 1/8"
  • Ideal for use with flexible tubing
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.