John Guest Push-To-Connect Fitting, Tube-to-Hose Elbow, Acetal, 3/8" OD×1/4" ID; 10/Pk
Supplier: VWR International
VWR Item#MFLX34006-85
Your Organization's Price:
Retrieving
No need to buy two fitting halves.
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.<br /><br />Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.