Masterflex® Fitting, Polypropylene, Y Hosebarb Reducer, 3/16" ID x 3/16" ID x 5/16" ID; 10/PK
Supplier: VWR International
VWR Item#MFLX30726-52
Your Organization's Price:
Retrieving
Ensure a tight seal every time.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/16" ID x 3/16" ID x 5/16" ID
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).