Masterflex® Fitting, HDPE, Straight, Hose Barb Reducer, 3/8" ID x 1/4" ID; 10/PK
Supplier: VWR International
VWR Item#MFLX30622-35
Your Organization's Price:
Retrieving
Best for low-pressure applications.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -65 to 150 °F (-53 to 65 °C)
  • Connection Type: 3/8" ID x 1/4" ID