Masterflex® Sanitary Gasket, Silicone, 3/4" Tri-Clamp®; 10/PK
Supplier: RUBBERFAB GASKET & MOLDING
VWR Item#MFLX30548-00
Your Organization's Price:
Retrieving
Ensure a tight seal.
  • Single-use custom configuration solutions to minimize downtime
  • Material: Silicone
  • Clamp Size: 3/4"
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.