Masterflex® Fitting, HDPE, Tee, Hose Barb Reducer, 1/2" ID x 1/2" ID x 3/8" ID; 100/PK
Supplier: VWR International
VWR Item#MFLX30401-54
Your Organization's Price:
Retrieving
Quick and easy assembly.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: ¹/₂" int.Ø × ¹/₂" int.Ø × ³/₈" int.Ø
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).