Heavy-Duty Tubing Pinch Clamp, Polysulfone
Supplier: VWR International
Durable design handles higher working pressures.
- Quick installation with positive swing-away locking screw
- Smooth contours allow for easy cleaning
Use this clamp with tubing or hose - it will meet the working pressure of either. High-strength polysulfone closure provides excellent durability, can be continually reused, and resists most chemicals. Simply align the hose within the clamp; mating surfaces on the clamp indicate complete closure. Typical applications include sterile filling, media bag connections, pumping, liquid control and high-purity fluid transfers. FDA-compliant polysulfone material can be sterilized by autoclave or gamma radiation.