Masterflex® L/S® Peristaltic Pump Replacement Brushes; 2/PK
Supplier: Avantor Fluid Handling
Quickly and easily replace components of your Masterflex system.
- Choose replacement brushes and caps based on your Masterflex L/S drive
- Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
Suitable for use with Masterflex® L/S series 07520, 07521, 07522, 07523, 07525, 07526, 07527, 07528, 07553 (except -50, -60, -90), 07554 (except -50, -60), 07558, 07567, 07568, 07592, 77300 drives.