Masterflex® I/P® Standard Pump Head, Replacement Rotor Assembly, Short Shaft, CRS Rotor
Supplier: Avantor Fluid Handling
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Constructed of durable cold-rolled steel (CRS)
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.