Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
Supplier: BEL-ART PRODUCTS
Deliver accurate regulation or interruption of fluid flow.
- Constructed of Delrin acetal
DELRIN® resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.