Masterflex® Hose Clamp, Acetal Copolymer, White, 0.928" ID x 1.072" ID; 100/PK
Supplier: VWR International
Reusable white acetal copolymer clamps will not rust or corrode.
- Resists corrosion
- Clamp size: 0.928" int.Ø × 1.072" int.Ø
Recommended for continuous use from ambient temperature to 200ºF (93ºC). Clamps are nonconducting and stable in a wide variety of chemicals. <br /><br />Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.
The acetal hose clamp kit includes 170 assorted clamps from 0.228" to 1.405" int.Ø.