Masterflex® Transfer Tubing, Ultramicrobore PTFE, 0.002" ID x 0.014" OD; 100 ft
Supplier: VWR International
VWR Item#MFLX06417-70
Your Organization's Price:
Retrieving
Minimize friction, maximize chemical resistance.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 0.002" ID x 0.014" OD
Use this translucent tubing in extreme temperatures and in harsh chemical environments. It's chemically inert, has low permeability, and has the lowest friction coefficient of any tubing. Temperature range is -400 to 500 °F (-240 to 260 °C); melts at 620 °F (327 °C). Tensile strength is 2500 to 4000 psi. PTFE tubing resin is UL recognized and complies with FDA (21 CFR 177.1550) and USP Class VI requirements.