Masterflex® Stainless Steel (SS) Hose Clamps, 3/4" x 1-3/4"; 10/Pk
Supplier: VWR International
VWR Item#MFLX06403-13
Your Organization's Price:
Retrieving
Ideal for securely clamping tubes in place.
  • Resists corrosion
  • Clamp size: 13/16" ID x 1-3/4" ID
The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.