Pentair Size 4 high-flow polyester filter bag; 300F (149C) max, 10 um porosity
Supplier: Avantor
Wide range of options for temperature, flow rate, and filtration quality.
- Constructed of Polyester Felt
Designed for higher flow with an average efficiency at the rated pore size. Polyester felt is ideal for general-purpose filtration. The bags are glazed to minimize fiber migration. Choose polyester for higher temperature applications, rated for a temperature range of 300 °F (149 °C) maximum.