GF Signet 163.546.351 Type 546 Ball Valve, CPVC with seals, 3/8"
Supplier: BURT PROCESS EQUIPMENT
VWR Item#MFLX01371-08
Your Organization's Price:
Retrieving
Optimize safety with this true union ball valve.
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.