Wide-Range Relief Valve, PVC, 1/2" NPT(F)
Supplier: VWR International
VWR Item#MFLX01350-62
Your Organization's Price:
Retrieving
Prevent leaks with the emergency shutdown design.
  • Three-seal system vents liquid to warn of approaching maintenance
Valves with connections of 1/4" or 1/2" have a diaphragm made of Viton® and withstand a maximum inlet pressure of 210 psig. All other valves have a PTFE shaft design and withstand a maximum inlet pressure of 140 psig. The shaft design prevents emergency shutdown with a three-seal system that will vent liquid to warn of approaching maintenance. All valves are recommended for liquid applications.