Esteem® Stretchy Vinyl Examination Gloves with Neu-Thera®, Cardinal
Supplier: Avantor
89042-010CS 384.65 USD
89042-010 89042-014 89042-016 89042-008
Esteem® Stretchy Vinyl Examination Gloves with Neu-Thera®, Cardinal
Gloves
Stretchy, powder-free vinyl gloves offer exceptional durability, dexterity, and a unique formulation that protects hands from moisture loss.
  • Beaded cuff
  • Ambidextrous
  • Smooth finish
  • Thickness: 4.1 mil (finger)
Latex-free gloves are ideal for short-term use (<15 minutes) where there is minimal stress, low risk, and low exposure to blood or other potentially infectious materials. Clinical tests have shown the Neu-Thera formulation to moisturize and soothe skin through dermal conditioning during wear.
Ordering information: Packaged 100 gloves per dispenser box, except x-large gloves (89042-016), which are packaged 90 gloves per dispenser box.