Personna® Double Edge Blades in Safety Case, AccuTec Blades, Inc.
Supplier: Avantor
76353-735CS 1029.45 USD
76353-735
Personna® Double Edge Blades in Safety Case, AccuTec Blades, Inc.
Razors and Blades
Double edge stainless steel blades in a safety case that is made to protect you and your products. Breakthrough workstation tool that speeds production while minimizing the risk of cuts.
  • Easy access safety case
  • Positive cap closure
  • Degreased blade for reduced oil and particulates
  • 3-facet blade edge for increased sharpness
  • High quality stainless steel
Eliminates potential nicks from unwrapping blades.