CoreSafe® Yellow Natural Rubber Chemical-Resistant Gloves, Flock-Lined, SW Safety Solutions
Supplier: Avantor
76199-990BG 20.18 USD
76199-990 76199-992 76199-994
CoreSafe® Yellow Natural Rubber Chemical-Resistant Gloves, Flock-Lined, SW Safety Solutions
Gloves
Chemical-resistant natural rubber gloves offer 13 mil of protection at the palm in a 13" length.
  • Yellow natural rubber latex
  • Fish scale grip pattern
  • Flock-lined for donning and comfort
  • Straight cut cuff
  • Imprinted cuff
  • Double chlorinated inner and outer surfaces
Flock-lined for comfort and ease of donning and doffing, these hand-specific, unsupported gloves with a fish scale grip pattern deliver the right combination of latex protection and performance.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; EAC TP TC 019/2011; CE 0493.
Ordering information: Glove sizes are as follows; S (7), M (8), L (9), XL (10).
Packaging: Gloves are packaged 12 pairs per bag (144 pairs per case).