PowerForm® PF-106-110-TTK/BAL-GR/YW Nitrile Exam Gloves with TracTek® Performance Grip, SW Safety Solutions
Supplier: Avantor
76051-170BX 28.8 USD
76051-170 76051-172 76051-174 76051-176 76051-226
PowerForm® PF-106-110-TTK/BAL-GR/YW Nitrile Exam Gloves with TracTek® Performance Grip, SW Safety Solutions
Gloves
PowerForm® gloves work with confidence in rugged wet and oily environments with TracTek® and Breach Alert™ protection.
  • TracTek® Grip Technology is engineered to maximize surface contact and channel away liquids
  • Two-ply Breach Alert™ Visual Detection Technology provides the enhanced durability of a glove engineered with two distinct polymer layers
  • Extended cuff for added splash protection
  • Silicone-free for critical environments
  • AQL of 1.5 exceeds laboratory standards
PowerForm® brings new hand protection technologies to workplace performance. From sweat management to enhanced grip design, it features enable workers to execute tasks with more confidence. It focuses on functional advancements to optimize on-the-job performance every day.
The PowerForm® with TracTek® Grip Technology is the result of an extensive R and D effort aimed at achieving a new level of grip performance in wet and oily conditions. Featuring a high-density, pyramidal pattern that provides 270% more contact area than conventional grip gloves, the PowerForm® delivers on its' promise of maximum grip in the most demanding environments. Loaded with unique skin health and performance features like two-ply Breach Alert™ construction for durability and visual breach detection.
For decades SW® has been the premium manufacturer of single-use performance products. With a relentless focus on new manufacturing technologies and materials that drive innovation, SW® single-use styles continuously push the boundaries of quality and worker-enhancing performance that fuels business success. The SW® difference: 100% air inspected, 75% less residue, consistent tip to cuff.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; ANSI Puncture Level 2; EAC TP TC 019/2011; CE 0493; PowerForm® is Dermatalogically Approved by the Skin Health Alliance (SHA), Meets stringent standards that ensure skin safety.
Ordering information: Glove sizes are as follows; S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11), XXXL (11.5 to 12).