Potassium chlorate ≥99.0%, crystals ACS

Danger

CA71008-044EA 41.08 USD
CA71008-044
Potassium chlorate ≥99.0%, crystals ACS
Potassium chlorate
Formula: KClO₃
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 356 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 13,07703

Order Now

Specification Test Results

Assay (KClO3) 99.0% min.
Insoluble Matter 0.005% max.
Bromate (BrO3) 0.015% max.
Chloride (Cl) 0.001% max.
Sulfate (SO4) Passes Test
Heavy metals ( as Pb) 5 ppm max.
Iron (Fe) 3 ppm max.
Calcium (Ca) 0.002% max.
Magnesium (Mg) 0.002% max.
Sodium (Na) 0.01% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR