Water, Molecular Biology Grade

Supplier: VWR


Filtered through a 0.2 µm filter.

02-0201-1000 02-0201-0500
VWRL0201-1000CS 159.73 USD
VWRL0201-1000 VWRL0201-0500
Water, Molecular Biology Grade
Water

VWR® Molecular Biology Grade Water is 0.2 um filtered, free of DEPC (Diethyl pyrocarbonate) and other chemical additives. The product is extremely low in endotoxin and has been analyzed for the absence of DNase, RNase and Protease. It is suitable in molecular biology research applications.


  • Low endotoxin
  • Free of DEPC
  • DNase, RNase and Protease free


Applications:
Reagent/Media/Buffer preparation
Molecular biology applications
DNA and RNA applications
Protein applications
PCR
NGS
QC and/or analytical applications
Wet Chemistry
LC-MS


Caution: For research and further manufacturing use. Not for Therapeutic Applications.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Conductivity before packaging ≤ 1.3 uS/cm
DNase (EQC) None
Endotoxin (EQC) ≤ 0.25 EU/ml
Microorganism Growth (EQC) No Growth
pH (EQC) Passes test
Protease (EQC) None
RNase (EQC) None
Total Organic Carbon < 500ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR