VWR® Trypsin (0.25%) EDTA (1X)
Supplier: VWR
02-0154-0100
VWRL0154-0100CS 1341.93 USD
VWRL0154-0100
VWR® Trypsin (0.25%) EDTA (1X)
Enzymes
Trypsin (0.25%) EDTA (1X) is used to release adherent cells from tissue culture plasticware for harvesting and passaging.
  • Used to release adherent cells
  • Aids in harvesting and passaging
Contains 0.001% phenol red, 0.25% Trypsin, 0.04% EDTA.