VWR® Hank's Balanced Salt Solution (HBSS)
Supplier: VWR
02-0121-0500
VWRL0121-0500CS 249.91 USD
VWRL0121-0500
VWR® Hank's Balanced Salt Solution (HBSS)
Buffers Cell Culture Buffers
Hanks' Balanced Salt Solution (HBSS) is a buffer used to maintain a physiological pH for cells maintained in non-CO2 atmospheric conditions.
  • HBSS can be used for washing cells
  • HBSS can be used as a solution for transporting cells and tissue culture