VWR® Fluorometer

Supplier: VWR International
VWRI210-000157
VWRI210-000157EA 3100 USD
VWRI210-000157
VWR® Fluorometer
Spectrophotometers and Photometers Fluorescence Spectrophotometers

VWR® Fluorometer for quantitation of DNA, RNA, and Proteins. Compact with touch screen control panel. Compatible with VWR® quantitation kits and kits from leading suppliers. Excitation: Blue 460 to 480 nm, Red 630 to 650 nm; Emission: Green 500 to 535 nm, Red 670 to 710 nm. 100 to 240 VAC.



  • Sensitive quantitation down to 0.1 pg/µl ds DNA

  • Large color touch screen for programming and operation
  • Two models for single or eight-sample processing
  • Built-in reagent calculators for sample preparation
  • Compact and portable (11×6" footprint)


Sample volume: 1 to 20 µl
Compatible sample tubes: 0.5 ml transparent thin wall PCR tube
Linearity range: Five orders of magnitude
Sensitivity: dsDNA: 0.01 ng/µl, ssDNA: 0.05 ng/µl, RNA: 0.25 ng/µl, microRNA: 0.05 ng/µl
Calibration: 2 or 3 points
Control panel: 5" color touch screen
Working environment: 15 to 30 ℃, <75% RH, indoor use



Accessories information:

VWR® High Sensitivity dsDNA Quantification kit provides an accurate method for quantitation of dsDNA samples when sensitivity is required. Highly selective for dsDNA vs RNA. Kit includes concentrated fluorescent reagent, dilution buffer and dsDNA standards. Detection range of 0.1 to 120 ng.

VWR® 1X dsDNA High sensitivity quantification kit is designed specifically for efficient and accurate quantitation of dsDNA. Premixed working solution saves on prep time. The kit is highly selective for dsDNA over RNA, and offers excellent stability, linear dynamic range, and sensitivity. Kit includes premixed fluorescent solution and dsDNA standards. Detection range of 0.1 to 120 ng.

VWR® Broad Range dsDNA Quantification kit provides an accurate method for quantitation of dsDNA samples for a broad range quantity of DNA. Highly selective for dsDNA vs RNA. Kit includes concentrated fluorescent reagent, dilution buffer and dsDNA standards. Detection range of 4 to 2000 ng.

VWR® 1X dsDNA Broad range quantification kit is designed specifically for efficient and accurate quantitation of dsDNA. Premixed working solution saves on prep time. The kit is highly selective for dsDNA over RNA, and offers excellent stability, linear dynamic range, and sensitivity. Kit includes premixed fluorescent solution and dsDNA standards. Detection range of 4 to 2000 ng.

VWR® ssDNA Quantification kit is designed for quantitation of oligonucleotides or long ssDNA. Contaminating proteins and nucleotides are not detected and will not interfere with results. The ssDNA kit also tolerates contamination of salts, solvents and detergents. Kit includes concentrated fluorescent reagent, dilution buffer and ssDNA standards. Detection range 1 to 200 ng.

VWR® High Sensitivity RNA quantification kit is highly selective for RNA in the presence of DNA and tolerate common contaminants such as free nucleotides, salts, solvents, detergents, and proteins. Kit includes concentrated fluorescent reagent, dilution buffer and RNA standards. This kit offers a detection range of 4 to 200 ng.

VWR® Broad Range RNA quantification kit is highly selective for RNA in the presence of DNA and tolerate common contaminants such as free nucleotides, salts, solvents, detergents, and proteins. Kit includes concentrated fluorescent reagent, dilution buffer and RNA standards. This kit offers a detection range of 10 to 1200 ng.

Optically clear micro tubes have a total volume capacity of 0.5 ml, and include a snap-close top.  Compatible with the VWR® Fluorometer and other popular fluorometer instruments.


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR