TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Bioreagent

Supplier: VWR

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

VWRB85827-1KG VWRB85827-5KG
VWRB85827-1KGEA 612.09 USD
VWRB85827-1KG VWRB85827-25KG VWRB85827-5KG VWRB85827-12KG
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Bioreagent
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)

Material is of non-animal, non-human origin

Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD00012590
CAS Number: 1185-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance white crystals or crystalline powder
Assay (By titration) NLT 99.0%
Absorbance (1M solution) 260nm NMT 0.05
Absorbance (1M solution) 280nm NMT 0.05
Absorbance (1M solution) 400nm NMT 0.01
Identification (by IR) conforms to structure
Insoluble Matter NMT 0.005%
Loss On Drying (at 105ºC) NMT 0.5%
Melting Point 147⁰C - 153⁰C
pH (05M sloution in H2O) 3.5 to 5.0
pKa (@ 25ºC) 8.0 to 8.4
Residue On Ignition NMT 0.1%
Solubility (1M solution in H2O) clear and complete
Arsenic (As) NMT 10ppm
Calcium (Ca) NMT 5 ppm
Copper (Cu) NMT 5 ppm
Iron (Fe) NMT 5 ppm
Lead (Pb) NMT 5 ppm
Magnesium (Mg) NMT 5 ppm
Manganese (Mn) NMT 5 ppm
Zinc (Zn) NMT 5 ppm
Bioburden NMT 100 CFU/g
Cell Culture Toxicity Passes test
Endotoxin (10% solution in H2O) NMT 0.1 EU/mg
DNase none detected
RNase none detected
Protease none detected

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR