Disodium hydrogen phosphate dihydrate Ph. Eur., USP, BP endotoxin tested

Supplier: Avantor Performance Materials

VWRB709-12KG
VWRB709-12KGEA 1251.04 USD
VWRB709-12KG VWRB709-1KG
Disodium hydrogen phosphate dihydrate Ph. Eur., USP, BP endotoxin tested
di-Sodium hydrogen phosphate dihydrate

Tested according to the USP, EP, and BP monographs. Material is of non-animal, non-human origin.


  • Bioprocessing
  • Suitable for use as an Excipient

Formula: Na₂HPO₄·2H₂O
Melting Pt: 92.5 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7

Order Now

Specification Test Results

Arsenic ≤ 16 ppm
Chloride ≤ 0.06 %
Bacterial Endotoxins ≤ 2.5 EU/g
Heavy Metals 231 ≤ 20 ppm
Identification (Phosphate) PASS
Identification (Sodium) PASS
Insoluble Substances ≤ 0.4 %
Loss on Drying 731 18.5 – 21.5%
Purity (dried basis) 98.0 - 100.5%
Sulfate ≤0.2 %
Appearance of solution PASS
Arsenic (2.4.2 Method A) ≤4 ppm
Chlorides ≤400 ppm
Heavy Metals (2.4.8) ≤ 20 ppm
Identification A PASS
Identification B PASS
Identification C PASS
Identification D PASS
Iron ≤40 ppm
Loss on Drying (2.2.32) 19.5 – 21.0%
Monosodium Phosphate ≤2.5%
Purity (dried substance) 98.0-101.0%
Reducing Substances PASS
Sulfates ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR