Calcium chloride dihydrate 99.0-107.0% USP, JP, Ph. Eur., BP suitable for use as excipient endotoxin tested

Supplier: Avantor Performance Materials

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

VWRB556-12KG VWRB556-1KG
VWRB556-12KGEA 823.71 USD
VWRB556-12KG VWRB556-1KG
Calcium chloride dihydrate 99.0-107.0% USP, JP, Ph. Eur., BP suitable for use as excipient endotoxin tested
Calcium chloride dihydrate

Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available.

Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176
Density: 1.85 g/cm³ (20 °C)
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay (USP) 99.0 – 107.0%
Endotoxin (USP) ≤ 2.5EU/g
Heavy Metals 231 (USP) ≤ 10ppm
Identification A (USP) PASS
Identification B (USP) PASS
Iron, Aluminum, and Phosphate (USP) PASS
Limit of Magnesium and Alkali Salts (USP) ≤ 1.0%
pH (50mg/mL, Water) @ 25C (USP) 4.5 – 9.2
Acidity or Alkalinity (EP & BP) PASS
Aluminum Test (EP & BP) PASS
Appearance of Solution (EP & BP) PASS
Assay (EP) (EP & BP) 97.0 – 103.0%
Barium Test (EP & BP) PASS
Heavy Metals (2.4.8) (EP & BP) ≤ 20ppm
Identification A (EP & BP) PASS
Identification B (EP & BP) PASS
Identification C (EP & BP) PASS
Iron (2.4.9) (EP & BP) ≤ 10ppm
Magnesium and Alkali Metals (EP & BP) ≤ 0.5%
Sulfates (2.4.13) (EP & BP) ≤ 300ppm
Arsenic 1.11 (JP) ≤ 2ppm
Assay (JP) 96.7% - 103.3%
Barium Test (JP) PASS
Clarity and Color of Solution (JP) PASS
Heavy Metals 1.07 (JP) ≤ 10ppm
Hypochlorite (JP) PASS
Identification (JP) PASS
Iron, Aluminum or Phosphate Test (JP) PASS
pH 2.54 (JP) 4.5 – 9.2
Sulfate 1.14 (JP) ≤ 0.024%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR