Sodium carbonate monohydrate Ph. Eur., NF, BP endotoxin tested

Supplier: Avantor Performance Materials
Warning

VWRB427-12KG
VWRB427-12KGEA 2097.74 USD
VWRB427-12KG
Sodium carbonate monohydrate Ph. Eur., NF, BP endotoxin tested
Sodium carbonate monohydrate

Tested according to the NF, EP, and BP monographs. Material is of non-animal, non-human origin.


  • Bioprocessing
  • Suitable for use as an Excipient

Formula: Na₂CO₃·1H₂O
Melting Pt: 107.85 °C
Density: 2.25 g/cm³ (20 °C)
MDL Number: MFCD00149177
CAS Number: 5968-11-6

Order Now

Specification Test Results

Assay (NF) 99.5 – 100.5%
Bacterial Endotoxins (NF) ≤ 2.5 EU/g
Heavy Metals 231 ≤ 10 ppm
Identification A (NF) PASS
Identification B (NF) PASS
Water Determination 12.0 – 15.0%
Alkali Hydroxides and Bicarbonates PASS
Appearance of solution PASS
Arsenic (2.4.2 Method A) ≤5 ppm
Assay 83.0 – 87.5%
Chlorides (2.4.4) ≤125 ppm
Heavy Metals (2.4.8) ≤50 ppm
Identification A PASS
Identification B PASS
Identification C PASS
Iron (2.4.9) ≤50 ppm
Sulfates (2.4.13) ≤250 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR