BD Vacutainer® Specialty Venous Blood Collection Tubes, BD Diagnostics
Supplier: BD
362753 362760 362761 364606 364960 364816 366703
BD362753CS 1125.42 USD
BD362753 BD362760 BD362761 VT4606 BD366703 VT369741 BD367855 VT4960 VT4816 BD-366703 BD366704
BD Vacutainer® Specialty Venous Blood Collection Tubes, BD Diagnostics
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Sterile vacuum blood collection tubes for speciality applications.
Tubes with a BD Hemogard™ closure incorporate a plastic shield that protects lab personnel from contact with blood on the stopper or around the outer rim of the tube, and from potential splattering when tube is opened. The closure's rubber stopper is recessed inside the plastic shield to isolate blood drops left by the needle.