BD Microtainer® Tubes, BD Diagnostics
Supplier: BD
365978 365963 365987 365992 365967 365974 365985 365965
VT365978CS 348.81 USD
VT365978 VT365963 BD365987 BD365992 VT365967 VT365974 VT365985 BD365965
BD Microtainer® Tubes, BD Diagnostics
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
BD Microtainer® tubes are designed for the collection, transport, and processing of skin puncture blood.
The “twist-assist” BD Microgard™ closure is ready-to-use and reduces blood splatter upon removal.
The integrated collection scoop, along with a wider diameter tube, allow for ease of collection and improved mixing. To simplify additive and sample identification, Microtainer® tubes are color-coded in a manner similar to evacuated tubes. Volume level lines, printed on tubes, ensure proper blood-to-anticoagulant ratio.