BD Vacutainer® Needle Holders, BD Biosciences
Supplier: BD
364815
VT364815CS 122.91 USD
VT364815
BD Vacutainer® Needle Holders, BD Biosciences
Blood Collection Tube Holders
Standard plastic holders accept disposable and reusable double-ended needles with threaded hubs
Holders convert BD Vacutainer® tube and needle into a syringe-like assembly that facilitates use. Not autoclavable.
Ordering information: To use needle holders for pediatric work, order adapter VT4597.