Water Fitting, WaterSaver Faucet
Supplier: WaterSaver Faucet
LE2800-131WS
VFT-0306-01EA 173.12 USD
VFT-0306-01
Water Fitting, WaterSaver Faucet
Faucets and Sinks Faucets
Curb- or panel-mounted chrome-plated water fitting consists of a single outlet turret and a straight water stop.
Ordering information: Complete with 9.5mm (3/8") IPS tank nipple, locknut, and washer.