Laboratory Ball Valve, WaterSaver Faucet
Supplier: WaterSaver Faucet
L4200-158WS
VFL4200-158WSEA 99.44 USD
VFL4200-158WS
Laboratory Ball Valve, WaterSaver Faucet
Faucets and Sinks Faucets
For installation on vertical or horizontal surfaces (such as pipe chases and service carriers) where there is access behind surface during installation.
  • Forged brass valve body
  • Forged brass lever handle with color-coded index disc
Inlet furnished with 3/8" IPS mounting shank (assembled), locknut and washer. 3/8" NPT male inlet. 3/8" NPT female outlet with removable ten serration hose end.