VWR® Fume Hoods
Supplier: VWR International
VFH5448-LN VFH5448-SW VFH5460-SW VFH5472-SW VFH5496-SW VFH5460-LN VFH5472-LN VFH5496-LN
VFH5448-LNEA 6745 USD
VFH5448-LN VFH5448-SW VFH5460-SW VFH5472-SW VFH5496-SW VFH5460-LN VFH5472-LN VFH5496-LN
VWR® Fume Hoods
Fume Hoods and Enclosures Chemical Fume Hoods
The VWR® fume hood is a general purpose fume hood, flexible and ready for the numerous applications called for in the modern laboratory.
  • Ready for standard or ADA configurations
  • Total of nine non-ADA fixture cut outs (plugged)
  • Total of three ADA compliant fixture cut outs (plugged)
  • Two airflow monitor cut outs
  • 120 V GFI duplex, with additional cut out for another duplex
  • Stainless steel airfoil
  • LED lighting
The VWR® fume hood has been methodically designed to provide end users a single fume hood that meets the numerous applications needed in the laboratory setting. Ranging from numerous fixture locations, available power, and air flow monitor cut outs, this fume hood can easily be converted to an ADA compliant fume hood.
Certifications:
ASHRAE 110, UL 1805.
Ordering information:
Work surface not included, must be ordered separately.
Delivery information:
Ships assembled and palletised.
Packaging:
The fume hood is palletised and packaged with high strength cardboard.
Caution:
It is recommended that a certified installer vetted by the VWR furniture team be utilised for proper installation.