Mini Fan, Nederman
Supplier: Nederman
70332961
VF332961EA 1407.45 USD
VF332961
Mini Fan, Nederman
Fume Extractors
The Mini Fan offers a 175cfm capacity, with 12.7cm (5") inlet and outlet ports
This fan is not equipped with a speed controller.