GRAVITYFLO® Gravity-Fed Eye/Face Wash, Speakman®
Supplier: Speakman
SE-4000
VF-4000-SEEA 2224.07 USD
VF-4000-SE
GRAVITYFLO® Gravity-Fed Eye/Face Wash, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
This portable, gravity-fed eye/face wash is perfect for environmental site locations as well as areas that are not plumbed.
  • Easy-carry molded hand grips are located in the water head
  • Can be wall mounted with a bracket (included) or shelf mounted
Constructed of durable high density plastic, this 60.6 L (16 gal.) unit features large rubber spray heads for complete eye and face coverage. Open the quick-acting valve by pulling the spray arms down. In the 'off' position, spray heads are enclosed in a recessed area for protection from contamination.
Certifications: Meets ANSI Z358-1 standard for portable eye wash units. Meets ANSI A117.1 standard for access and use by physically handicapped people.